missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDX56 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5148292
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
DDX56 Polyclonal Antibody for Western Blot, ICC/IF
This gene encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The protein encoded by this gene shows ATPase activity in the presence of polynucleotides and associates with nucleoplasmic 65S preribosomal particles. This gene may be involved in ribosome synthesis, most likely during assembly of the large 60S ribosomal subunit.
Tekniske data
| DDX56 | |
| Polyclonal | |
| Unconjugated | |
| DDX56 | |
| 2600001H07Rik; 61-kd nucleolar helicase; ATP-dependent 61 kDa nucleolar RNA helicase; D11Ertd619e; DDX21; DDX26; DDX56; DEAD (Asp-Glu-Ala-Asp) box helicase 56; DEAD (Asp-Glu-Ala-Asp) box polypeptide 56; DEAD box protein 21; DEAD box protein 56; DEAD-box helicase 56; DEAD-box RNA helicase; NOH61; nucleolar helicase of 61 kDa; nucleolar RNA helicase homolog; probable ATP-dependent RNA helicase DDX56; putative nucleolar RNA helicase; RH-II/Gu; RNA helicase II/Gu | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 54606 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 100 μg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q9NY93 | |
| DDX56 | |
| E. coli-derived Human DDX56 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion