missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX56, Mouse anti-Human, Clone: 4C5, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00054606-M05.100ug
This item is not returnable.
View return policy
Description
Sequence: IKEELLHSEKLKTYFEDNPRDLQLLRHDLPLHPAVVKPHLGHVPDYLVPPALRGLVRPHKKRKKLSSSCRKAKRAKSQNPLRSFKHKGKKFRPTAKPSSpecifications
| DDX56 | |
| Monoclonal | |
| Unconjugated | |
| DEAD (Asp-Glu-Ala-Asp) box polypeptide 56 | |
| DDX21/DDX26/NOH61 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 4C5 | |
| In 1x PBS, pH 7.4 | |
| NM_019082 | |
| DDX56 | |
| DDX56 (NP_061955, 450 a.a. ∼ 547 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 54606 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction