missing translation for 'onlineSavingsMsg'
Learn More

DDX56 Antibody, Novus Biologicals™

Product Code. 30228181 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30228181 100 μL 100µL
30230973 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30228181 Supplier Novus Biologicals Supplier No. NBP338610100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DDX56 Polyclonal antibody specifically detects DDX56 in Human,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX56
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, ELISA
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias ATP-dependent 61 kDa nucleolar RNA helicase, DDX26, DEAD (Asp-Glu-Ala-Asp) box polypeptide 56, DEAD box protein 21, DEAD box protein 56, DEAD-box RNA helicase, EC 3.6.1, EC 3.6.4.13,61-kd nucleolar helicase, NOH61DDX21, probable ATP-dependent RNA helicase DDX56, putative nucleolar RNA helicase
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 318-547 of human DDX56 (NP_061955.1).,, Sequence:, EVLGAPVKGKRRGRGPKGDKASDPEAGVARGIDFHHVSAVLNFDLPPTPEAYIHRAGRTARANNPGIVLTFVLPTEQFHLGKIEELLSGENRGPILLPYQFRMEEIEGFRYRCRDAMRSVTKQAIREARLKEIKEELLHSEKLKTYFEDNPRDLQLLRHDLPLHPAVVKPHLGHVPDYLVPPALRGLVRPHKKRKKLSSSCRKAKRAKSQNPLRSFKHKGKKFRPTAKPS
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 54606
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.