missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX56 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-97523CL1
This item is not returnable.
View return policy
Description
DDX56 Polyclonal antibody specifically detects DDX56 in Human samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| DDX56 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| ATP-dependent 61 kDa nucleolar RNA helicase, DDX26, DEAD (Asp-Glu-Ala-Asp) box polypeptide 56, DEAD box protein 21, DEAD box protein 56, DEAD-box RNA helicase, EC 3.6.1, EC 3.6.4.13,61-kd nucleolar helicase, NOH61DDX21, probable ATP-dependent RNA helicase DDX56, putative nucleolar RNA helicase | |
| Produced in rabbits immunized with E. coli-derived Human DDX56 fragment. | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 54606 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction