missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX55 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£130.00 - £290.00
Specifications
| Antigen | DDX55 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18339141
|
Bio-Techne
NBP3-15971-20UL |
20 μg |
£130.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18399926
|
Novus Biologicals
NBP3-15971-100UL |
100 μg |
£290.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DDX55 Polyclonal antibody specifically detects DDX55 in Human, Mouse samples. It is validated for Western BlotSpecifications
| DDX55 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, 50% glycerol, pH7.3 | |
| 57696 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| DEAD (Asp-Glu-Ala-Asp) box polypeptide 55, DEAD box protein 55, EC 3.6.1, EC 3.6.4.13, FLJ16577, KIAA1595ATP-dependent RNA helicase DDX55, MGC33209 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-360 of human DDX55 (NP_065987.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title