missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDX55 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536431
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 68 kDa. Purity is >95% by SDS-PAGE.
This gene encodes an enzyme which ubiquitinates proteins which participate in signaling pathways and apoptosis.
Specifications
| DDX55 | |
| Polyclonal | |
| Unconjugated | |
| DDX55 | |
| 2810021H22Rik; ATP-dependent RNA helicase DDX55; Ddx55; DEAD (Asp-Glu-Ala-Asp) box polypeptide 55; DEAD box protein 55; DEAD-box helicase 55; Kiaa1595; mKIAA1595 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57696 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NHQ9 | |
| DDX55 | |
| Synthetic peptide corresponding to amino acids 102-150 of human DDX55. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction