missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX54 Mouse anti-Human, Clone: 1H9F4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66664-1-IG-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The nucleolar protein encoded by this gene interacts in a hormone-dependent manner with nuclear receptors, and represses their transcriptional activity. Alternative splice variants that encode different isoforms have been found for this gene.Specifications
| DDX54 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8TDD1 | |
| DDX54 | |
| DDX54 Fusion Protein Ag25289 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 1H9F4 | |
| Unconjugated | |
| DDX54 | |
| DDX54, DEAD box protein 54, DEAD box RNA helicase 97 kDa, DP97 | |
| Mouse | |
| Protein A | |
| RUO | |
| 79039 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction