missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX54 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-89028-25ul
This item is not returnable.
View return policy
Description
DDX54 Polyclonal specifically detects DDX54 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| DDX54 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| APR-5, ATP-dependent RNA helicase DP97, DEAD (Asp-Glu-Ala-Asp) box polypeptide 54, DEAD box helicase 97 KDa, DEAD box protein 54, DEAD box RNA helicase 97 kDa, DP97ATP-dependent RNA helicase DDX54, EC 3.6.1, EC 3.6.4.13, MGC2835 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| DDX54 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:VDTKLNEQLKTSFFLVREDTKAAVLLHLLHNVVRPQDQTVVFVATKHHAEYLTELLTTQRVSCAHIYSALDPTARKINLAKFTLGKCSTLIVTDLAARGLDIPLLDNVINYSFP | |
| 25 μL | |
| Signal Transduction | |
| 79039 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction