missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX54 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-60678
This item is not returnable.
View return policy
Description
DDX54 Polyclonal specifically detects DDX54 in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot (Negative).
Specifications
| DDX54 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| APR-5, ATP-dependent RNA helicase DP97, DEAD (Asp-Glu-Ala-Asp) box polypeptide 54, DEAD box helicase 97 KDa, DEAD box protein 54, DEAD box RNA helicase 97 kDa, DP97ATP-dependent RNA helicase DDX54, EC 3.6.1, EC 3.6.4.13, MGC2835 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500 | |
| NP_076977.3 | |
| DDX54 | |
| The immunogen recognized by this antibody maps to a region between residue 800 and 850 of human DEAD (Asp-Glu-Ala-Asp) box polypeptide 54 using the numbering given in entry NP_076977.3 (GeneID 79039). | |
| 100 μL | |
| Signal Transduction | |
| 79039 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction