missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DDX53 Monoclonal Antibody (CL7899)
GREENER_CHOICE

Product Code. 30827859
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30827859 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30827859 Supplier Invitrogen™ Supplier No. MA562959
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

DDX53 Monoclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX53
Applications Western Blot
Classification Monoclonal
Clone CL7899
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DDX53
Gene Accession No. Q86TM3
Gene Alias CAGE; cancer/testis antigen 26; Cancer-associated gene protein; CT26; DDX53; DEAD (Asp-Glu-Ala-Asp) box polypeptide 53; DEAD box protein 53; DEAD box protein CAGE; DEAD-box helicase 53; DEAD-box protein CAGE; probable ATP-dependent RNA helicase DDX53
Gene Symbols DDX53
Host Species Mouse
Immunogen Recombinant protein corresponding to Human DDX53.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 168400
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.