Learn More
DDX53 Antibody (CL7899) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-88909JF669
Product Code. 30307419
  • £558.00 / 0.10mL
Description
DDX53 Monoclonal antibody specifically detects DDX53 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DDX53
Monoclonal
Janelia Fluor 669
CAGEprobable ATP-dependent RNA helicase DDX53, Cancer/testis antigen 26, Cancer-associated gene protein, CT26EC 3.6.4.13, DEAD (Asp-Glu-Ala-Asp) box polypeptide 53, DEAD box protein 53, DEAD box protein CAGE, DEAD-box protein CAGE, EC 3.6.1
This antibody was developed using a synthetic peptide derived from Q86TM3, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer
168400
Store at 4⪚C in the dark.
IgG1
Western Blot
CL7899
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified