missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDX52 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563257
This item is not returnable.
View return policy
Description
Immunogen sequence: KFDTRRFSAD AARFQIGKRK YDFDSSEVLQ GLDFFGNKKS VPGVCGASQT HQKPQNGEKK EESLTERKRE QSKKKRKTMT SEIAS Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 60%.
The product encoded by this gene is a member of a RNA-binding protein family and possesses nucleolytic activity against cytotoxic lymphocyte (CTL) target cells. It has been suggested that this protein may be involved in the induction of apoptosis as it preferentially recognizes poly(A) homopolymers and induces DNA fragmentation in CTL targets. The major granule-associated species is a 15-kDa protein that is thought to be derived from the carboxyl terminus of the 40-kDa product by proteolytic processing. Alternative splicing resulting in different isoforms of this gene product has been described in the literature.
Specifications
| DDX52 | |
| Polyclonal | |
| Unconjugated | |
| DDX52 | |
| 2700029C06Rik; ATP-dependent RNA helicase ROK1-like; ATP-dependent, RNA helicase; DDX52; DEAD (Asp-Glu-Ala-Asp) box polypeptide 52; DEAD box protein 52; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 52; DEAD-box helicase 52; HUSSY19; HUSSY-19; probable ATP-dependent RNA helicase DDX52; Rok1; rROK1L | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11056 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2R4 | |
| DDX52 | |
| Recombinant protein corresponding to Human DDX52. Recombinant protein control fragment (Product #RP-93272). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction