missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX52 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71811
This item is not returnable.
View return policy
Description
DDX52 Polyclonal specifically detects DDX52 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| DDX52 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| ATP-dependent RNA helicase ROK1-like, DEAD (Asp-Glu-Ala-Asp) box polypeptide 52, DEAD box protein 52, EC 3.6.1, HUSSY19, probable ATP-dependent RNA helicase DDX52, ROK1EC 3.6.4.13 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 11056 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
| NP_008941.2 | |
| DDX52 | |
| The immunogen for this product maps to a region between residue 549 and 599 of human DEAD (Asp-Glu-Ala-Asp) Box Polypeptide 52 using the numbering given in entry NP_008941.2 (GeneID 11056). | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction