missing translation for 'onlineSavingsMsg'
Learn More

DDX52 Antibody, DyLight 650, Novus Biologicals™

Product Code. 18415614 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18415614 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18415614 Supplier Novus Biologicals Supplier No. NBP171810C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DDX52 Polyclonal antibody specifically detects DDX52 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX52
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias ATP-dependent RNA helicase ROK1-like, DEAD (Asp-Glu-Ala-Asp) box polypeptide 52, DEAD box protein 52, EC 3.6.1, HUSSY19, probable ATP-dependent RNA helicase DDX52, ROK1EC 3.6.4.13
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 499 and 549 of human DEAD (Asp-Glu-Ala-Asp) Box Polypeptide 52 using the numbering given in entry NP_008941.2 (GeneID 11056).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11056
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.