missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX51 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20149-1-AP-20UL
This item is not returnable.
View return policy
Description
Involved in nucleolar processing of pre-18S ribosomal RNA.Specifications
| DDX51 | |
| Polyclonal | |
| Unconjugated | |
| DDX51 | |
| DDX51, DEAD box protein 51 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 304570, 317781, 69663 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6P9R1, Q8N8A6 | |
| DDX51 | |
| DDX51 Fusion Protein Ag13691 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction