missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX47 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35128-20ul
This item is not returnable.
View return policy
Description
DDX47 Polyclonal antibody specifically detects DDX47 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| DDX47 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| DEAD (Asp-Glu-Ala-Asp) box polypeptide 47, DEAD box polypeptide 47, DEAD box protein 47, DKFZp564O176, E4-DBP, E4-DEAD box protein, EC 3.6.1, EC 3.6.4.13, FLJ30012, HQ0256, MSTP162, probable ATP-dependent RNA helicase DDX47, RRP3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 186-455 of human DDX47 (NP_057439.2).,, Sequence:, TEVDKILKVIPRDRKTFLFSATMTKKVQKLQRAALKNPVKCAVSSKYQTVEKLQQYYIFIPSKFKDTYLVYILNELAGNSFMIFCSTCNNTQRTALLLRNLGFTAIPLHGQMSQSKRLGSLNKFKAKARSILLATDVASRGLDIPHVDVVVNFDIPTHSKDYIHRVGRTARAGRSGKAITFVTQYDVELFQRIEHLIGKKLPGFPTQDDEVMMLTERVAEAQRFARMELREHGEKKKRSREDAGDNDDTEGAIGVRNKVAGGKMKKRKGR | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 51202 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction