missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDX46 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530335
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (99%), Rat (96%), Pig (99%), Rhesus Monkey (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The protein encoded by this gene is a component of the 17S U2 snRNP complex; it plays an important role in pre-mRNA splicing.
Specifications
| DDX46 | |
| Polyclonal | |
| Unconjugated | |
| DDX46 | |
| 2200005K02Rik; 8430438J23Rik; AI325430; AI957095; DDX46; DEAD (Asp-Glu-Ala-Asp) box polypeptide 46; DEAD box polypeptide 46; DEAD box protein 46; DEAD-box helicase 46; Hel117; Helicase of 117.4 kDa; Kiaa0801; mKIAA0801; Probable ATP-dependent RNA helicase DDX46; Prp5; PRP5 homolog; Prp5-like DEAD-box protein; PRPF5; RNA helicase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9879 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q7L014 | |
| DDX46 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human DDX46. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction