missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DDX43 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA556722

Product Code. 15724631

  • £452.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: GIDTAFQPSV GKDGSTDNNV VAGDRPLIDW DQIREEGLKW QKTKWADLPP IKKNFYKEST ATSAMSKVEA DSWR Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 65%.

The protein encoded by this gene is an ATP-dependent RNA helicase in the DEAD-box family and displays tumor-specific expression.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DDX43
Polyclonal
Unconjugated
DDX43
Cancer/testis antigen 13; CT13; DDX43; DEAD (Asp-Glu-Ala-Asp) box polypeptide 43; DEAD box protein 43; DEAD box protein HAGE; DEAD-box helicase 43; DEAD-box protein 43; ENSMUSG00000070291; HAGE; Helical antigen; OTTMUSG00000019690; probable ATP-dependent RNA helicase DDX43
Rabbit
Antigen Affinity Chromatography
RUO
55510
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9NXZ2
DDX43
Recombinant protein corresponding to Human DDX43. Recombinant protein control fragment (Product #RP-94633).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.