missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDX43 Polyclonal Antibody

Description
The antibody detects endogenous levels of total DDX43 protein.
The protein encoded by this gene is an ATP-dependent RNA helicase in the DEAD-box family and displays tumor-specific expression.
Specifications
Specifications
| Antigen | DDX43 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | DDX43 |
| Gene Accession No. | Q9NXZ2 |
| Gene Alias | Cancer/testis antigen 13; CT13; DDX43; DEAD (Asp-Glu-Ala-Asp) box polypeptide 43; DEAD box protein 43; DEAD box protein HAGE; DEAD-box helicase 43; DEAD-box protein 43; ENSMUSG00000070291; HAGE; Helical antigen; OTTMUSG00000019690; probable ATP-dependent RNA helicase DDX43 |
| Gene Symbols | DDX43 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?