missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX43 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-28701AF594
This item is not returnable.
View return policy
Description
DDX43 Polyclonal antibody specifically detects DDX43 in Human samples. It is validated for Western Blot
Specifications
| DDX43 | |
| Polyclonal | |
| Western Blot | |
| Cancer/testis antigen 13, CT13DEAD-box protein 43, DEAD (Asp-Glu-Ala-Asp) box polypeptide 43, DEAD box protein 43, DKFZp434H2114, EC 3.6.1, EC 3.6.4.13, HAGEDEAD box protein HAGE, Helical antigen, probable ATP-dependent RNA helicase DDX43 | |
| The immunogen recognized by this antibody maps to a region between residue 275 and 325 of human DEAD (Asp-Glu-Ala-Asp) box polypeptide 43 using the numbering given in entry NP_061135.1 (GeneID 55510). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 55510 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction