missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDX42 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546803
This item is not returnable.
View return policy
Description
Peptide sequence: SREGTGGSNG KRERYTENRG SSRHSHGETG NRHSDSPRHG DGGRHGDGYR.
This gene encodes a member of the Asp-Glu-Ala-Asp (DEAD) box protein family. Members of this protein family are putative RNA helicases, and are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. Two transcript variants encoding the same protein have been identified for this gene.
Specifications
| DDX42 | |
| Polyclonal | |
| Unconjugated | |
| DDX42 | |
| 1810047H21Rik; ATP-dependent RNA helicase DDX42; AW319508; AW556242; B430002H05Rik; Ddx42; DDX42P; DEAD (Asp-Glu-Ala-Asp) box helicase 42; DEAD (Asp-Glu-Ala-Asp) box polypeptide 42; DEAD box protein 42; DEAD-box helicase 42; RHELP; RNA helicase-like protein; RNA helicase-related protein; RNAHP; SF3b DEAD box protein; SF3b125; SF3B8; splicing factor 3b, subunit 8; splicing factor 3B-associated 125 kDa protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11325 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q86XP3 | |
| DDX42 | |
| Synthetic peptide directed towards the C-terminal of human DDX42 (aa 796-845). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction