missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DDX42 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30481
This item is not returnable.
View return policy
Description
DDX42 Polyclonal antibody specifically detects DDX42 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| DDX42 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 11325 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| DEAD (Asp-Glu-Ala-Asp) box polypeptide 42, DEAD box protein 42, EC 3.6.1, EC 3.6.4.13, RHELPSF3b DEAD box protein, RNA helicase-like protein, RNA helicase-related protein, RNAHPFLJ43179, SF3b125 DEAD-box protein, SF3b125ATP-dependent RNA helicase DDX42, Splicing factor 3B-associated 125 kDa protein | |
| between 800 and 850 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction