missing translation for 'onlineSavingsMsg'
Learn More

DDX42 Antibody, Novus Biologicals™

Product Code. 18382429 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18382429 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18382429 Supplier Novus Biologicals Supplier No. H00011325B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

DDX42 Polyclonal antibody specifically detects DDX42 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX42
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunocytochemistry/ Immunofluorescence
Formulation PBS (pH 7.4)
Gene Accession No. AAH15505.1
Gene Alias DEAD (Asp-Glu-Ala-Asp) box polypeptide 42, DEAD box protein 42, EC 3.6.1, EC 3.6.4.13, RHELPSF3b DEAD box protein, RNA helicase-like protein, RNA helicase-related protein, RNAHPFLJ43179, SF3b125 DEAD-box protein, SF3b125ATP-dependent RNA helicase DDX42, Splicing factor 3B-associated 125 kDa protein
Host Species Mouse
Immunogen DDX42 (AAH15505.1, 1 a.a. - 819 a.a.) full-length human protein. MAEVEDQAARDMKRLEEKDKERKNVKGIRDDIEEEDDQEAYFRYMAENPTAGVVQEEEEDNLEYDSDGNPIAPTKKIIDPLPPIDHSEIDYPPFEKNFYNEHEEITNLTPQQLIDLRHKLNLRVSGAAPPRPGSSFAHFGFDEQLMHQIRKSEYTQPTPIQCQGVPVALSGRDMIGIAKTGSGKTAAFIWPMLIHIMDQKELEPGDGPIAVIVCPTRELCQQIHAECKRFGKAYNLRSVAVYGGGSMWEQAKALQEGAEIVVCTPGRLIDHVKKKATNLQRVSYLVFDEADRMFDMGFEYQVRSIASHVRPDRQTLLFSATFRKKIEKLARDILIDPIRVVQGDIGEANEDVTQIVEILHSGPSKWNWLTRRLVEFTSSGSVLLFVTKKANAEELANNLKQEGHNLGLLHGDMDQSERNKVISDFKKKDIPVLVATDVAARGLDIPSIKTVINYDVARDIDTHTHRIGRTGRAGEKGVAYTLLTPKDSNFAGDLVRNLEGANQHVSKELLDLAMQNAWFRKSRFKGGKGKKLNIGGGGLGYRERPGLGSENMDRGNNNVMSNYEAYKPSTGAMGDRLTAMKAAFQSQYKSHFVAASLSNQKAGSSAAGASGWTSAGSLNSVPTNSAQQGHNSPDSPVTSAAKGIPGFGNTGNISGAPVTYPSAGAQGVNNTASGNNSREGTGGSNGKRERYTENRGSSRHSHGETGNRHSDSPRHGDGGRHGDGYRHPESSSRHTDGHRHGENRHGGSAGRHGENRGANDGRNGESRKEAFNRESKMEPKMEPKVDSSKMDKVDSKTDKTADGFAVPEPPKRKKSRWDS
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11325
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.