missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDX41 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529240
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, H1299, HepG2, Raji. Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (83%), Chicken (93%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Probable ATP-dependent RNA helicase DDX41 is a DEAD box protein characterized by the conserved motif Asp-Glu-Ala-Asp. It is required during post-transcriptional gene expression and may be involved in pre-mRNA splicing. Mutations affecting the gene can result in myeloproliferative/lymphoproliferative neoplasms, familial (MPLPF).
Specifications
| DDX41 | |
| Polyclonal | |
| Unconjugated | |
| DDX41 | |
| 2900024F02Rik; AA958953; ABS; Abstrakt; AI324246; Ddx41; DEAD (Asp-Glu-Ala-Asp) box polypeptide 41; DEAD box protein 41; DEAD box protein abstrakt homolog; DEAD-box helicase 41; DEAD-box protein abstrakt; MPLPF; probable ATP-dependent RNA helicase DDX41; putative RNA helicase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51428 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q9UJV9 | |
| DDX41 | |
| Recombinant protein encompassing a sequence within the center region of human DDX41. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto