Learn More
Invitrogen™ DDX41 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109625
Description
Immunogen sequence: MEESEPERKR ARTDEVPAGG SRSEAEDEDD EDYVPYVPLR QRRQLLLQKL LQRRRKGAAE EEQQDSGSEP RGDEDDIPLG PQSNVSLLDQ HQHLKEKAEA RKESAKEKQL KEEEKILESV AEGRALMSVK EMAKGITYDD PIKTSWTPPR YVLSMSEERH ERVRKKYHIL VEGDGIPPPI KSFKEMKFPA AILRGLKKKG IHHPTPIQIQ GIPTILSGRD MIGIAFTGSG KTLVFTLPVI MFCLEQEKRL PFSKREGPYG LIICPSRELA RQTHGILEYY CRLLQEDSSP LLRCALCIGG.
Probable ATP-dependent RNA helicase DDX41 is a DEAD box protein characterized by the conserved motif Asp-Glu-Ala-Asp. It is required during post-transcriptional gene expression and may be involved in pre-mRNA splicing. Mutations affecting the gene can result in myeloproliferative/lymphoproliferative neoplasms, familial (MPLPF).
Specifications
| DDX41 | |
| Polyclonal | |
| Unconjugated | |
| DDX41 | |
| 2900024F02Rik; AA958953; ABS; Abstrakt; AI324246; Ddx41; DEAD (Asp-Glu-Ala-Asp) box polypeptide 41; DEAD box protein 41; DEAD box protein abstrakt homolog; DEAD-box helicase 41; DEAD-box protein abstrakt; MPLPF; probable ATP-dependent RNA helicase DDX41; putative RNA helicase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 314336, 51428, 72935 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1.91 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q91VN6, Q9UJV9 | |
| DDX41 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human DDX41 (NP_057306.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.