Learn More
DDX4,VASA Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-51042-20UL
Description
The DDX4,VASA antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human DDX4,VASA. This antibody recognizes human, mouse antigen. The DDX4,VASA antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| DDX4,VASA | |
| 1000 μg/mL | |
| DDX4, DEAD box protein 4, VASA, Vasa homolog | |
| 20 μL | |
| 54514 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |