missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDX4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583387
This item is not returnable.
View return policy
Description
Immunogen sequence: GGESSDTQGP KVTYIPPPPP EDEDSIFAHY QTGINFDKYD TILVEVSGHD APPAILTFEE ANLCQTLNNN IAKAGYTK Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 99%.
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp, are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is a homolog of VASA proteins in Drosophila and several other species. The gene is specifically expressed in the germ cell lineage in both sexes and functions in germ cell development. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| DDX4 | |
| Polyclonal | |
| Unconjugated | |
| DDX4 | |
| ATP-dependent RNA helicase DDX4; AV206478; Cvh; Ddx4; D-E-A-D (aspartate-glutamate-alanine-aspartate) box polypeptide 4; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 4; DEAD (Asp-Glu-Ala-Asp) box polypeptide 4; DEAD box polypeptide 4; DEAD box protein 4; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 4; DEAD-box helicase 4; MGC111074; Mvh; mvh / m'vasa; probable ATP-dependent RNA helicase DDX4; putative ATP-dependent RNA helicase DDX4; RVLG; VASA; Vasa homolog; vasa-like gene protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54514 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NQI0 | |
| DDX4 | |
| Recombinant protein corresponding to Human DDX4. Recombinant protein control fragment (Product #RP-96902). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction