Learn More
Invitrogen™ DDX3Y Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522050
Description
Recommended positive controls: H1299, HepG2, mouse brain, rat testis, Jurkat, Raji, NCI-H929. Predicted reactivity: Mouse (95%), Rat (94%), Rhesus Monkey (99%), Chimpanzee (97%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases which are implicated in a number of cellular processes involving RNA binding and alteration of RNA secondary structure. DDX3Y is one member of DEAD box RNA helicases, encode by the gene locate on the chromosome Y in the azoospermia factor A region.
Specifications
| DDX3Y | |
| Polyclonal | |
| Unconjugated | |
| Ddx3y | |
| 8030469F12Rik; ATP-dependent RNA helicase DDX3Y; D1Pas1 related sequence 1; D1Pas1-related sequence 1; D1Pas1-rs1; DBY; Ddx3y; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide, Y chromosome; DEAD (Asp-Glu-Ala-Asp) box helicase 3, Y-linked; DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, Y-linked; DEAD box protein 3, Y-chromosomal; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide, Y chromosome; Dead2; DEAD-box helicase 3, Y-linked; DEAD-box RNA helicase DEAD2; mDEAD2; RGD1309586; uncharacterized protein LOC364073 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26900, 364073, 8653 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.89 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| O15523, Q62095 | |
| Ddx3y | |
| Recombinant protein encompassing a sequence within the center region of human DDX3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.