missing translation for 'onlineSavingsMsg'
Learn More

DDX39B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16823624 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16823624 20 μL 20µL
16813624 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16823624 Supplier Proteintech Supplier No. 147981AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody may cross-react with DDX39A.

This gene encodes a member of the DEAD box family of RNA-dependent ATPases that mediate ATP hydrolysis during pre-mRNA splicing. The encoded protein is an essential splicing factor required for association of U2 small nuclear ribonucleoprotein with pre-mRNA, and also plays an important role in mRNA export from the nucleus to the cytoplasm. A cluster of genes, BAT1-BAT5, is localized in the vicinity of the genes for TNF alpha and TNF beta. These genes are all within the human major histocompatibility complex class III region. Mutations in this gene may be associated with rheumatoid arthritis. Alternatively spliced transcript variants encoding the same protein have been described.

Specifications

Antigen DDX39B
Applications Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DDX39B
Gene Accession No. Q13838, Q63413, Q9Z1N5
Gene Alias ATP dependent RNA helicase p47, BAT1, D6S81E, DDX39B, DEAD box protein UAP56, HLA B associated transcript 1, Spliceosome RNA helicase BAT1, UAP56
Gene Symbols Ddx39b
Host Species Rabbit
Immunogen DDX39B Fusion Protein Ag6512
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114612, 53817, 7919
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.