missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX39 Polyclonal Antibody

DDX39 Polyclonal Antibody
Brand: Invitrogen PA576309
This item is not returnable.
View return policy
Description
DDX39 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP, ELISA
Specifications
| DDX39 | |
| Polyclonal | |
| Unconjugated | |
| DDX39A | |
| 2610307C23Rik; ATP-dependent RNA helicase DDX39; ATP-dependent RNA helicase DDX39A; BAT1; BAT1L; DDX39; Ddx39a; Ddxl; DEAD (Asp-Glu-Ala-Asp) box polypeptide 39; DEAD (Asp-Glu-Ala-Asp) box polypeptide 39A; DEAD box protein 39; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 39; DEAD-box helicase 39A; nuclear RNA helicase URH49; Nuclear RNA helicase, DECD variant of DEAD box family; UAP56-related helicase, 49 kDa; URH49 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10212, 68278, 89827 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O00148, Q5U216, Q8VDW0 | |
| Ddx39, DDX39A | |
| DDX39A fusion protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction