missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDX39 Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Rat (96%), Xenopus laevis (92%), Rhesus Monkey (100%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the DEAD box protein family. These proteins are characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD) and are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of the DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene is thought to play a role in the prognosis of patients with gastrointestinal stromal tumors. A pseudogene of this gene is present on chromosome 13. Alternate splicing results in multiple transcript variants. Additional alternatively spliced transcript variants of this gene have been described, but their full-length nature is not known.
Specifications
Specifications
| Antigen | DDX39 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | DDX39A |
| Gene Accession No. | O00148 |
| Gene Alias | 2610307C23Rik; ATP-dependent RNA helicase DDX39; ATP-dependent RNA helicase DDX39A; BAT1; BAT1L; DDX39; Ddx39a; Ddxl; DEAD (Asp-Glu-Ala-Asp) box polypeptide 39; DEAD (Asp-Glu-Ala-Asp) box polypeptide 39A; DEAD box protein 39; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 39; DEAD-box helicase 39A; nuclear RNA helicase URH49; Nuclear RNA helicase, DECD variant of DEAD box family; UAP56-related helicase, 49 kDa; URH49 |
| Gene Symbols | DDX39A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?