missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX39 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98622-50ul
This item is not returnable.
View return policy
Description
DDX39 Polyclonal antibody specifically detects DDX39 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| DDX39 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| ATP-dependent RNA helicase DDX39, BAT1, BAT1L, DDX39, DDXL, DEAD (Asp-Glu-Ala-Asp) box polypeptide 39, DEAD (Asp-Glu-Ala-Asp) box polypeptide 39 transcript, DEAD (Asp-Glu-Ala-Asp) box polypeptide 39A, DEAD box protein 39, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 39, EC 3.6.4.13, MGC18203, MGC8417, Nuclear RNA helicase URH49, nuclear RNA helicase, DECD variant of DEAD box family, UAP56-related helicase, 49 kDa, URH49 | |
| Produced in rabbits immunized with E. coli-derived Human DDX39 fragment. | |
| 50 μg | |
| DNA replication Transcription Translation and Splicing | |
| 10212 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction