missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX39 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £359.00
Specifications
| Antigen | DDX39 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18626140
|
Novus Biologicals
NBP2-92080-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18694271
|
Novus Biologicals
NBP2-92080-0.1ml |
0.1 mL |
£359.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DDX39 Polyclonal antibody specifically detects DDX39 in Mouse, Rat samples. It is validated for Western BlotSpecifications
| DDX39 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| DNA replication Transcription Translation and Splicing | |
| PBS with 50% glycerol, pH7.3. | |
| 10212 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| ATP-dependent RNA helicase DDX39, BAT1, BAT1L, DDX39, DDXL, DEAD (Asp-Glu-Ala-Asp) box polypeptide 39, DEAD (Asp-Glu-Ala-Asp) box polypeptide 39 transcript, DEAD (Asp-Glu-Ala-Asp) box polypeptide 39A, DEAD box protein 39, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 39, EC 3.6.4.13, MGC18203, MGC8417, Nuclear RNA helicase URH49, nuclear RNA helicase, DECD variant of DEAD box family, UAP56-related helicase, 49 kDa, URH49 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 178-427 of human DDX39A (NP_005795.2). LVRNRSFSLKNVKHFVLDECDKMLEQLDMRRDVQEIFRLTPHEKQCMMFSATLSKDIRPVCRKFMQDPMEVFVDDETKLTLHGLQQYYVKLKDSEKNRKLFDLLDVLEFNQVIIFVKSVQRCMALAQLLVEQNFPAIAIHRGMAQEERLSRYQQFKDFQRRILVATNLFGRGMDIERVNIVFNYDMPEDSDTYLHRVARAGRFGTKGLAITFVSDENDAKILNDVQDRFEVNVAELPEEIDISTYIEQSR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title