missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX3 Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 81903-1-RR-100UL
This item is not returnable.
View return policy
Beskrivelse
The DDX3 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human DDX3. This antibody recognizes human, mouse, rat antigen. The DDX3 antibody has been validated for the following applications: WB, IF/ICC, FC (Intra), ELISA analysis.Tekniske data
| DDX3 | |
| Recombinant Monoclonal | |
| 1000 μg/mL | |
| DBX; DDX14; DDX3; HLP2 | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 75 kDa | |
| Liquid |
| Western Blot, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA | |
| 5C12 | |
| Unconjugated | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 1654 | |
| Store at -20°C. Stable for one year after shipment. | |
| Recombinant Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion