missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDX28 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559780
This item is not returnable.
View return policy
Description
Immunogen sequence: TLLDESFLEL VDYILEKSHI AEGPADLEDP FNPKAQLVLV GATFPEGVGQ LLNKVASPDA VTTITSSKLH CIMPHVKQTF LRLKGADK Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 82%.
WDR11 is a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This gene is located in the chromosome 10q25-26 region, which is frequently deleted in gliomas and tumors of other tissues, and is disrupted by the t(10;19) translocation rearrangement in glioblastoma cells. The gene location suggests that it is a candidate gene for the tumor suppressor locus.
Specifications
| DDX28 | |
| Polyclonal | |
| Unconjugated | |
| DDX28 | |
| 2410004K13Rik; AI449652; ATP-dependent RNA helicase Ddx28; DDX28; DEAD (Asp-Glu-Ala-Asp) box polypeptide 28; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 28; DEAD-box helicase 28; MDDX28; Mitochondrial DEAD box protein 28; mitochondrial DEAD-box polypeptide 28; probable ATP-dependent RNA helicase DDX28 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55794 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NUL7 | |
| DDX28 | |
| Recombinant protein corresponding to Human DDX28. Recombinant protein control fragment (Product #RP-98780). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur