missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX28 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92376-0.1ml
This item is not returnable.
View return policy
Description
DDX28 Polyclonal antibody specifically detects DDX28 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| DDX28 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DEAD (Asp-Glu-Ala-Asp) box polypeptide 28, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 28, EC 3.6.1, EC 3.6.4.13, FLJ11282, M, MDDX28probable ATP-dependent RNA helicase DDX28, Mitochondrial DEAD box protein 28, mitochondrial DEAD-box polypeptide 28 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 371-540 of human DDX28 (NP_060850.2). RLKGADKVAELVHILKHRDRAERTGPSGTVLVFCNSSSTVNWLGYILDDHKIQHLRLQGQMPALMRVGIFQSFQKSSRDILLCTDIASRGLDSTGVELVVNYDFPPTLQDYIHRAGRVGRVGSEVPGTVISFVTHPWDVSLVQKIELAARRRRSLPGLASSVKEPLPQAT | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 55794 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction