missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX27 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17087-1-AP-20UL
This item is not returnable.
View return policy
Description
La is involved in diverse aspects of RNA metabolism, including binding and protecting 3-prime UUU(OH) elements of newly RNA polymerase III.Specifications
| DDX27 | |
| Polyclonal | |
| Unconjugated | |
| DDX27 | |
| DDX27, DEAD box protein 27, HSPC259, PP3241, RHLP, Rrp3p | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 228889, 362274, 55661 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunocytochemistry | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q921N6, Q96GQ7 | |
| Ddx27 | |
| DDX27 Fusion Protein Ag9426 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction