missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX27 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£378.00
Specifications
| Antigen | DDX27 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
DDX27 Polyclonal specifically detects DDX27 in Human samples. It is validated for Western Blot.Specifications
| DDX27 | |
| Polyclonal | |
| Purified | |
| RUO | |
| DEAD (Asp-Glu-Ala-Asp) box polypeptide 27, DEAD box protein 27, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 27, dJ686N3.1, DKFZp667N057, DRS1, EC 3.6.1, EC 3.6.4.13, FLJ12917, FLJ20596, FLJ22238, MGC1018, MGC163147, PP3241, probable ATP-dependent RNA helicase DDX27, RHLP, RNA helicase-like protein, Rrp3p | |
| DDX27 | |
| IgG | |
| Protein A purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Q96GQ7 | |
| 55661 | |
| Synthetic peptides corresponding to DDX27 (DEAD (Asp-Glu-Ala-Asp) box polypeptide 27) The peptide sequence was selected from the N terminal of DDX27. Peptide sequence DEKIEKVRKKRKTEDKEAKSGKLEKEKEAKEGSEPKEQEDLQENDEEGSE. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title