missing translation for 'onlineSavingsMsg'
Learn More

DDX27 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18432567 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432567 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18432567 Supplier Novus Biologicals Supplier No. NBP126601AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DDX27 Polyclonal antibody specifically detects DDX27 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX27
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DEAD (Asp-Glu-Ala-Asp) box polypeptide 27, DEAD box protein 27, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 27, dJ686N3.1, DKFZp667N057, DRS1, EC 3.6.1, EC 3.6.4.13, FLJ12917, FLJ20596, FLJ22238, MGC1018, MGC163147, PP3241, probable ATP-dependent RNA helicase DDX27, RHLP, RNA helicase-like protein, Rrp3p
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 700 and 750 of human DEAD (Asp-Glu-Ala-Asp) box polypeptide 27 using the numbering given in entry NP_060365.6 (GeneID 55661).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55661
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.