missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX25 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | DDX25 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
DDX25 Polyclonal specifically detects DDX25 in Human samples. It is validated for Western Blot.Specifications
| DDX25 | |
| Polyclonal | |
| Rabbit | |
| Stem Cell Markers | |
| DEAD (Asp-Glu-Ala-Asp) box polypeptide 25, DEAD box protein 25, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 25, EC 3.6.1, EC 3.6.4.13, Gonadotropin-regulated testicular RNA helicase, GRTHATP-dependent RNA helicase DDX25 | |
| DDX25 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q9UHL0-2 | |
| 29118 | |
| Synthetic peptides corresponding to DDX25 (DEAD (Asp-Glu-Ala-Asp) box polypeptide 25) The peptide sequence was selected from the C terminal of DDX25. Peptide sequence TVEMIQDGHQVSLLSGELTVEQRASIIQRFRDGKEKVLITTNVCARGIDV. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title