missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX25 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £358.00
Specifications
| Antigen | DDX25 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
DDX25 Polyclonal antibody specifically detects DDX25 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| DDX25 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Stem Cell Markers | |
| PBS with 50% glycerol, pH7.3. | |
| 29118 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DEAD (Asp-Glu-Ala-Asp) box polypeptide 25, DEAD box protein 25, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 25, EC 3.6.1, EC 3.6.4.13, Gonadotropin-regulated testicular RNA helicase, GRTHATP-dependent RNA helicase DDX25 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 284-483 of human DDX25 (NP_037396.3). DSVWHFAERIIPDPNVIKLRKEELTLNNIRQYYVLCEHRKDKYQALCNIYGSITIGQAIIFCQTRRNAKWLTVEMIQDGHQVSLLSGELTVEQRASIIQRFRDGKEKVLITTNVCARGIDVKQVTIVVNFDLPVKQGEEPDYETYLHRIGRTGRFGKKGLAFNMIEVDELPSLMKIQDHFNSSIKQLNAEDMDEIEKIDY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title