Learn More
DDX21 Mouse anti-Human, CoraLite 594, Clone: 1B9C8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-66925-100UL
Description
DDX21 protein belongs to DEAD box protein family which is characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD). As a putative RNA helicase, DDX21 unwinds double-stranded RNA, folds single-stranded RNA and is involved in process including ribosomal RNA biogeneis, RNA editing and general transcription. Interaction of DDX21 and c-Jun was reported in ribosomal RNA processing.
Specifications
| DDX21 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Immunofluorescence, Flow Cytometry | |
| 1B9C8 | |
| CoraLite 594 | |
| 100 μL | |
| 9188 | |
| Liquid |