missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX21 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB100-1718SS
This item is not returnable.
View return policy
Description
DDX21 Polyclonal specifically detects DDX21 in Human, Mouse, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation (ChIP).
Specifications
| DDX21 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| DEAD (Asp-Glu-Ala-Asp) box polypeptide 21, DEAD box protein 21, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 21, DKFZp686F21172, EC 3.6.1, EC 3.6.4.13, Gu protein, GUA, gu-alpha, GURDB, nucleolar RNA helicase 2, Nucleolar RNA helicase Gu, Nucleolar RNA helicase II, RH II/Gu, RH-II/GU, RH-II/GuA, RNA helicase II/Gu alpha | |
| Rabbit | |
| 87 kDa | |
| 0.025 mL | |
| Primary | |
| Human, Mouse, Zebrafish | |
| IgG |
| Western Blot, ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000-1:10000, Chromatin Immunoprecipitation reported in scientific literature (PMID 32231306), Flow Cytometry 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Immunoprecipitation 2 - 10 ug/mg of lysate, Immunohistochemistry-Paraffin 1:10-1:500, Chromatin Immunoprecipitation (ChIP), Knockdown Validated reported in scientific literature (PMID 35440492) | |
| Q9NR30 | |
| DDX21 | |
| The immunogen recognized by this antibody maps to a region between residue 725 and the C-terminus (residue 783) of human DEAD/H (Asp-Glu-Ala-Asp/His) Box Polypeptide 21 using the numbering given in entry NP_004719.2 (GeneID 9188). | |
| Affinity Purified | |
| RUO | |
| 9188 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction