missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DDX19B Polyclonal Antibody
GREENER_CHOICE

Product Code. 15574992
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15574992 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15574992 Supplier Invitrogen™ Supplier No. PA541737

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: DEQEAAAESL SNLHLKEEKI KPDTNGAVVK TNANAEKTDE EEKEDRAAQS Sequence homology: Cow: 93%; Dog: 79%; Guinea Pig: 81%; Horse: 100%; Human: 100%; Mouse: 92%; Pig: 100%; Rabbit: 86%; Rat: 100%.

Spectrins are a family of filamentous cytoskeletal proteins that function as essential scaffold proteins that stabilize the plasma membrane and organize intracellular organelles. Spectrins are composed of alpha and beta dimers that associate to form tetramers linked in a head-to-head arrangement. This gene encodes an alpha spectrin that is specifically expressed in nonerythrocytic cells. The encoded protein has been implicated in other cellular functions including DNA repair and cell cycle regulation. Mutations in this gene are the cause of early infantile epileptic encephalopathy-5. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX19B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DDX19B
Gene Accession No. Q9UMR2
Gene Alias 2810457M08Rik; 4921519L13Rik; ATP-dependent RNA helicase DDX19; ATP-dependent RNA helicase DDX19B; AW260119; DBP5; DDX19; Ddx19b; DEAD (Asp-Glu-Ala-As) box polypeptide 19B; DEAD (Asp-Glu-Ala-Asp) box polypeptide 19b; DEAD box protein 19B; DEAD box RNA helicase DEAD5; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 19 (Dbp5, yeast, homolog); DEAD-box helicase 19B; DEAD-box protein 5; DEAD-box RNA helicase DEAD5; RNAh; TDBP; Unknown (protein for MGC:134354); yeast Dbp5 homolog; Zd10a; zinc responsive protein Zd10A
Gene Symbols DDX19B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human DDX19B (aa 11-60).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11269
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.