missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX19A Antibody (OTI3C12), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-45044-30ul
This item is not returnable.
View return policy
Description
DDX19A Monoclonal antibody specifically detects DDX19A in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| DDX19A | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| 55308 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |
| Immunohistochemistry, Western Blot | |
| OTI3C12 | |
| Western Blot 1:500-2000, Immunohistochemistry 1:500 | |
| ATP-dependent RNA helicase DDX19A, DDX19-DDX19L, DDX19L, DDX19-like protein, DEAD (Asp-Glu-Ala-As) box polypeptide 19A, DEAD (Asp-Glu-Ala-As) box polypeptide 19-like, DEAD box protein 19A, DKFZp686C21137, EC 3.6.1, EC 3.6.4.13, FLJ11126, RNA helicase | |
| Full length human recombinant protein of human DDX19A (NP_060802) produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction