missing translation for 'onlineSavingsMsg'
Learn More

DDX19A Antibody (OTI3C12) [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30689200 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30689200 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30689200 Supplier Novus Biologicals Supplier No. NBP344850JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DDX19A Monoclonal antibody specifically detects DDX19A in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX19A
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone OTI3C12
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias ATP-dependent RNA helicase DDX19A, DDX19-DDX19L, DDX19L, DDX19-like protein, DEAD (Asp-Glu-Ala-As) box polypeptide 19A, DEAD (Asp-Glu-Ala-As) box polypeptide 19-like, DEAD box protein 19A, DKFZp686C21137, EC 3.6.1, EC 3.6.4.13, FLJ11126, RNA helicase
Host Species Mouse
Immunogen Full length human recombinant protein of human DDX19A (NP_060802) produced in HEK293T cell.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55308
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.