missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DDX18 Polyclonal specifically detects DDX18 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | DDX18 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | DEAD (Asp-Glu-Ala-Asp) box polypeptide 18, DEAD box protein 18, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 18 (Myc-regulated), EC 3.6.1, EC 3.6.4.13, FLJ33908, MrDbATP-dependent RNA helicase DDX18, Myc-regulated DEAD box protein |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the C terminal region of human DDX18 (NP_006764.3). Peptide sequence VALSFGFKVPPFVDLNVNSNEGKQKKRGGGGGFGYQKTKKVEKSKIFKHI |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?