missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDX18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA523463
This item is not returnable.
View return policy
Description
DDX18 Polyclonal Antibody for Western Blot, IHC (P)
ATP-dependent RNA helicase DDX18 belongs to theDEAD box helicase family, which contains one helicase ATP-binding domain and one helicase C-terminal domain. DDX18 is induced by Myc and Max and is involved in splicing and nuclear export. It is reported to be differentially expressed during viral latency and at early times after induction into active replication.
Specifications
| DDX18 | |
| Polyclonal | |
| Unconjugated | |
| DDX18 | |
| ATP-dependent RNA helicase DDX18; cPERP-D; DDX18; DEAD (Asp-Glu-Ala-Asp) box polypeptide 18; DEAD box protein 18; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 18 (Myc-regulated); DEAD-box helicase 18; MrDb; Myc-regulated DEAD box protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 8886 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.05% BSA and 0.05% sodium azide | |
| Q9NVP1 | |
| DDX18 | |
| A portion of amino acids 20-80 of human DDX18. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction