missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX18 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£156.00 - £301.00
Specifications
| Antigen | DDX18 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 2-4ug/ml∼, Immunohistochemistry-Paraffin 5-10ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18171402
|
Novus Biologicals
NBP2-27353SS |
0.025 mg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18119232
|
Novus Biologicals
NBP2-27353 |
0.1 mg |
£301.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DDX18 Polyclonal specifically detects DDX18 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| DDX18 | |
| Western Blot 2-4ug/ml∼, Immunohistochemistry-Paraffin 5-10ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| DEAD (Asp-Glu-Ala-Asp) box polypeptide 18, DEAD box protein 18, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 18 (Myc-regulated), EC 3.6.1, EC 3.6.4.13, FLJ33908, MrDbATP-dependent RNA helicase DDX18, Myc-regulated DEAD box protein | |
| DDX18 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen for the DDX18 antibody is 100% homologous in human and 64% homologous in mouse. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 8886 | |
| A portion of amino acids 20-80 of human DDX18 was used as the immunogen for the antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title