missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDX17 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541913
This item is not returnable.
View return policy
Description
Peptide sequence: TSSANNPNLM YQDECDRRLR GVKDGGRRDS ASYRDRSETD RAGYANGSGY Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 100%.
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and splicesosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is an ATPase activated by a variety of RNA species, but not by dsDNA. This protein, and that encoded by DDX5 gene, are more closely related to each other than to any other member of the DEAD box family. This gene can encode multiple isoforms due to both alternative splicing and the use of alternative translation initiation codons, including a non-AUG (CUG) start codon.
Specifications
| DDX17 | |
| Polyclonal | |
| Unconjugated | |
| DDX17 | |
| 2610007K22Rik; A430025E01Rik; AI047725; C80929; DDX17; DEAD (Asp-Glu-Ala-Asp) box helicase 17; DEAD (Asp-Glu-Ala-Asp) box polypeptide 17; DEAD (Asp-Glu-Ala-Asp) box polypeptide 46; DEAD box polypeptide 17; DEAD box protein 17; DEAD box protein p72; DEAD box protein p82; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 17 (72kD); DEAD/H box 17; DEAD-box helicase 17; DKFZp761H2016; Gm926; P72; Probable ATP-dependent RNA helicase DDX17; RH70; RNA-dependent helicase p72; RP3-434P1.1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10521 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q92841 | |
| DDX17 | |
| Synthetic peptide directed towards the N-terminal of human DDX17 (aa 570-619). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction